Assay Details
Target Gene Details
Entrez Gene ID: | 11318 |
Gene Name: | G protein-coupled receptor 182 |
Gene Aliases: |
7TMR, ADMR, AM-R, AMR, G10D, gamrh, hrhAMR |
Location: |
Chr.12:56993895-56998441 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR182 | NM_007264.3 | 2 | 357 | NP_009195.1 |
| XM_011537824.2 | 2 | 1033 | XP_011536126.1 | |
| XM_011537825.2 | 2 | 1033 | XP_011536127.1 | |
| XM_011537827.2 | 2 | 1033 | XP_011536129.1 | |
| BC034761.1 | 2 | 482 | AAH34761.1 | |
| Y13583.1 | 2 | 357 | CAA73910.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| esv3580285 | Chr.12:56993106 - 57003042 on Build GRCh38 | Loss |
|
| nsv832428 | Chr.12:56946825 - 57141911 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map