Assay Details
Target Gene Details
Entrez Gene ID: | 26985 |
Gene Name: | adaptor related protein complex 3 mu 1 subunit |
Gene Aliases: |
- |
Location: |
Chr.10:74120257-74151085 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP3M1 | NM_001320263.1 | 6 | 1130 | NP_001307192.1 |
| NM_001320264.1 | 4 | 969 | NP_001307193.1 | |
| NM_001320265.1 | 4 | 681 | NP_001307194.1 | |
| NM_012095.5 | 4 | 843 | NP_036227.1 | |
| NM_207012.3 | 5 | 969 | NP_996895.1 | |
| NR_135191.1 | 4 | 843 | ||
| AF092092.1 | 3 | 509 | AAD20446.1 | |
| AK026983.1 | 4 | 578 | BAB15614.1 | |
| AK299352.1 | 4 | 432 | ||
| AK315597.1 | 4 | 552 | ||
| AL833820.1 | 2 | 148 | CAD38682.1 | |
| BC026232.1 | 5 | 728 | AAH26232.1 | |
| BC067127.1 | 4 | 939 | AAH67127.1 | |
| BU172918.1 | 4 | 807 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551527 | Chr.10:73949148 - 74140704 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map