Assay Details
Target Gene Details
Entrez Gene ID: | 91461 |
Gene Name: | protein kinase domain containing, cytoplasmic |
Gene Aliases: |
SGK493, Vlk |
Location: |
Chr.2:42048021-42058528 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKDCC | NM_138370.2 | NP_612379.2 | ||
| BC007901.2 | AAH07901.2 | |||
| BC062988.1 | AAH62988.1 | |||
| BC094697.1 | AAH94697.1 | |||
| BC110513.1 | AAI10514.1 | |||
| BC110514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953772 | Chr.2:42046161 - 42061760 on Build GRCh38 | Deletion |
|
| nsv833903 | Chr.2:41952790 - 42134588 on Build GRCh38 | Gain |
|
| nsv1005527 | Chr.2:42026606 - 42078931 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map