Assay Details
Target Gene Details
Entrez Gene ID: | 221301 |
Gene Name: | family with sequence similarity 26 member D |
Gene Aliases: |
C6orf78 |
Location: |
Chr.6:116529013-116561123 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM26D | NM_001256887.2 | 4 | 1455 | NP_001243816.1 |
| NM_001256888.2 | 4 | 1361 | NP_001243817.1 | |
| NM_001256889.2 | 3 | 1239 | NP_001243818.1 | |
| NM_153036.4 | 3 | 1294 | NP_694581.1 | |
| XM_005266860.3 | 2 | 2229 | XP_005266917.1 | |
| XM_011535560.2 | 5 | 1549 | XP_011533862.1 | |
| XM_011535561.2 | 4 | 1398 | XP_011533863.1 | |
| XM_011535562.2 | 6 | 1730 | XP_011533864.1 | |
| XM_011535563.2 | 4 | 1343 | XP_011533865.1 | |
| XM_011535564.1 | 4 | 1304 | XP_011533866.1 | |
| XM_017010390.1 | 5 | 1477 | XP_016865879.1 | |
| XM_017010391.1 | 5 | 1432 | XP_016865880.1 | |
| XM_017010392.1 | 5 | 1494 | XP_016865881.1 | |
| AK056801.1 | BAB71288.1 | |||
| AY358252.1 | AAQ88619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830783 | Chr.6:116445706 - 116645069 on Build GRCh38 | Gain |
|
| nsv523972 | Chr.6:115958024 - 116809541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map