Assay Details
Target Gene Details
Entrez Gene ID: | 7571 |
Gene Name: | zinc finger protein 23 |
Gene Aliases: |
KOX16, ZNF359, ZNF612, Zfp612 |
Location: |
Chr.16:71447597-71462252 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF23 | NM_001304492.1 | 5 | 606 | NP_001291421.1 |
| NM_001304493.1 | 7 | 731 | NP_001291422.1 | |
| NM_001304494.1 | 3 | 324 | NP_001291423.1 | |
| NM_145911.2 | 6 | 1178 | NP_666016.1 | |
| AF507946.1 | 6 | 1169 | AAM33359.1 | |
| AK091039.1 | 5 | 601 | ||
| AK297130.1 | 3 | 324 | ||
| AL080123.1 | 1 | 208 | CAB45722.1 | |
| AL833815.1 | 2 | 948 | CAD38678.1 | |
| BC007256.1 | 6 | 1169 | AAH07256.1 | |
| BC048974.1 | 7 | 725 | AAH48974.1 | |
| BT007400.1 | 3 | 355 | AAP36064.1 | |
| BX537507.1 | 1 | 2980 | CAD97769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064435 | Chr.16:71406816 - 71454868 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map