Assay Details
Target Gene Details
Entrez Gene ID: | 254359 |
Gene Name: | zinc finger DHHC-type containing 24 |
Gene Aliases: |
- |
Location: |
Chr.11:66520625-66546200 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZDHHC24 | NM_207340.1 | 3 | 1134 | NP_997223.1 |
| XM_005273874.3 | XP_005273931.1 | |||
| XM_011544894.1 | XP_011543196.1 | |||
| AK301787.1 | 3 | 1168 | ||
| AK310561.1 | ||||
| AY358448.1 | 3 | 1134 | AAQ88813.1 | |
| BC005015.1 | 2 | 548 | AAH05015.1 | |
| BC057833.1 | 3 | 963 | AAH57833.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map