Assay Details
Target Gene Details
Entrez Gene ID: | 820 |
Gene Name: | cathelicidin antimicrobial peptide |
Gene Aliases: |
CAP-18, CAP18, CRAMP, FALL-39, FALL39, HSD26, LL37 |
Location: |
Chr.3:48223347-48225491 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMP | NM_004345.4 | 1 | 255 | NP_004336.3 |
| AY162210.1 | 1 | 90 | AAN78318.1 | |
| AY251531.1 | 1 | 93 | AAP20054.1 | |
| BC055089.1 | 1 | 132 | AAH55089.1 | |
| BU074012.1 | 1 | 258 | ||
| CR457083.1 | 1 | 90 | CAG33364.1 | |
| CR541961.1 | 1 | 90 | CAG46759.1 | |
| U19970.1 | 1 | 105 | AAA74084.1 | |
| X89658.1 | 1 | 125 | CAA61805.1 | |
| Z38026.1 | 1 | 101 | CAA86115.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834682 | Chr.3:48096647 - 48269632 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map