Assay Details
Target Gene Details
Entrez Gene ID: | 139596 |
Gene Name: | uracil phosphoribosyltransferase homolog |
Gene Aliases: |
FUR1, UPP |
Location: |
Chr.X:75274059-75304897 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UPRT | NM_001307944.1 | 7 | 1475 | NP_001294873.1 |
| NM_145052.3 | 7 | 1425 | NP_659489.1 | |
| NR_030774.1 | 8 | 1483 | ||
| AK056354.1 | 7 | 1444 | ||
| AY643714.1 | 7 | 1411 | ||
| BC015116.1 | 7 | 1360 | AAH15116.1 | |
| CR749336.1 | 8 | 1399 | CAH18190.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25282 | Chr.X:75242143 - 75437229 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map