Assay Details
Target Gene Details
Entrez Gene ID: | 283985 |
Gene Name: | fatty acid desaturase 6 |
Gene Aliases: |
FP18279 |
Location: |
Chr.17:74874793-74893811 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FADS6 | NM_178128.5 | 6 | 1244 | NP_835229.3 |
| XM_005257224.4 | 6 | 1238 | XP_005257281.2 | |
| XM_017024458.1 | 7 | 1019 | XP_016879947.1 | |
| AK094411.1 | 6 | 1092 | BAC04349.1 | |
| AK293720.1 | 3 | 603 | ||
| AY203929.1 | 6 | 1053 | AAP34452.1 | |
| BC117230.1 | 6 | 1073 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
|
| esv3554779 | Chr.17:74876872 - 74881870 on Build GRCh38 | Deletion |
|
| nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
| esv3641248 | Chr.17:74877348 - 74881649 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map