Assay Details
Target Gene Details
Entrez Gene ID: | 84630 |
Gene Name: | tau tubulin kinase 1 |
Gene Aliases: |
BDTK |
Location: |
Chr.6:43243484-43288259 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTBK1 | NM_032538.1 | NP_115927.1 | ||
| XM_011514947.2 | XP_011513249.1 | |||
| XM_017011364.1 | XP_016866853.1 | |||
| XM_017011365.1 | XP_016866854.1 | |||
| XM_017011366.1 | XP_016866855.1 | |||
| XM_017011367.1 | XP_016866856.1 | |||
| AB058758.1 | BAB47484.1 | |||
| AB218664.1 | BAE78660.1 | |||
| AK131217.1 | BAD18405.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv602997 | Chr.6:43221202 - 43291349 on Build GRCh38 | Loss |
|
| nsv602998 | Chr.6:43245739 - 43308652 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map