Assay Details
Target Gene Details
Entrez Gene ID: | 6152 |
Gene Name: | ribosomal protein L24 |
Gene Aliases: |
HEL-S-310, L24 |
Location: |
Chr.3:101681090-101686719 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL24 | NM_000986.3 | NP_000977.1 | ||
| AK311992.1 | ||||
| BC000690.2 | AAH00690.1 | |||
| BC070193.1 | AAH70193.1 | |||
| BU754101.1 | ||||
| CD513036.1 | ||||
| CR456729.1 | CAG33010.1 | |||
| EU668341.1 | ||||
| M94314.1 | AAC28251.1 |
More Information
Set Membership: |
|

Genomic Map