Assay Details
Target Gene Details
Entrez Gene ID: | 51322 |
Gene Name: | WW domain containing adaptor with coiled-coil |
Gene Aliases: |
BM-016, DESSH, PRO1741, Wwp4 |
Location: |
Chr.10:28532426-28623112 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WAC | NM_100264.2 | 1 | 319 | NP_567822.1 |
| XM_011519491.2 | 1 | 481 | XP_011517793.1 | |
| XM_017016315.1 | 1 | 139 | XP_016871804.1 | |
| XM_017016316.1 | 1 | 481 | XP_016871805.1 | |
| XM_017016317.1 | 1 | 139 | XP_016871806.1 | |
| XM_017016318.1 | 1 | 139 | XP_016871807.1 | |
| AK055852.1 | 1 | 136 | BAB71029.1 | |
| AK225846.1 | 1 | 132 | ||
| BI463901.1 | 1 | 419 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734606 | Chr.10:27447765 - 29432048 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map