Assay Details
Target Gene Details
Entrez Gene ID: | 85377 |
Gene Name: | MICAL like 1 |
Gene Aliases: |
MICAL-L1, MIRAB13 |
Location: |
Chr.22:37906148-37942458 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MICALL1 | NM_033386.3 | NP_203744.1 | ||
| XM_005261791.3 | XP_005261848.1 | |||
| XM_005261792.3 | XP_005261849.1 | |||
| XM_011530471.2 | XP_011528773.1 | |||
| XM_011530472.2 | XP_011528774.1 | |||
| XM_011530473.2 | XP_011528775.1 | |||
| XM_011530474.2 | XP_011528776.1 | |||
| XM_011530475.2 | XP_011528777.1 | |||
| XM_011530476.2 | XP_011528778.1 | |||
| XM_017029026.1 | XP_016884515.1 | |||
| AJ496196.1 | CAD42713.1 | |||
| AL834373.1 | CAD39036.1 | |||
| CB997694.1 | ||||
| CR456437.1 | CAG30323.1 | |||
| DR004306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893482 | Chr.22:37832559 - 37927500 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map