Assay Details
Target Gene Details
Entrez Gene ID: | 84823 |
Gene Name: | lamin B2 |
Gene Aliases: |
EPM9, LAMB2, LMN2 |
Location: |
Chr.19:2428165-2456968 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMNB2 | NM_032737.3 | 12 | 4093 | NP_116126.3 |
| AK129641.1 | 2 | 1028 | ||
| M94362.1 | 11 | 3743 | AAA80979.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160562 | Chr.19:2328434 - 2460264 on Build GRCh38 | Deletion |
|
| nsv1064930 | Chr.19:2380093 - 2592252 on Build GRCh38 | Gain |
|
| nsv953944 | Chr.19:2283702 - 2525802 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map