Assay Details
Target Gene Details
Entrez Gene ID: | 56914 |
Gene Name: | otoraplin |
Gene Aliases: |
FDP, MIAL1 |
Location: |
Chr.20:16748353-16752164 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTOR | NM_020157.3 | 4 | 823 | NP_064542.1 |
| AF233261.1 | 5 | 765 | AAF82078.1 | |
| AF243505.1 | 4 | 781 | AAG42356.1 | |
| AJ242552.1 | 4 | 774 | CAC27443.1 | |
| BI495573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv90n6 | Chr.20:16750430 - 16759338 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map