Assay Details
Target Gene Details
Entrez Gene ID: | 79629 |
Gene Name: | occludin/ELL domain containing 1 |
Gene Aliases: |
FWP009, S863-9 |
Location: |
Chr.19:17226204-17229219 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OCEL1 | NM_024578.2 | NP_078854.1 | ||
| XM_005260079.2 | XP_005260136.1 | |||
| XM_006722899.3 | XP_006722962.1 | |||
| XM_017027306.1 | XP_016882795.1 | |||
| AF109191.1 | AAQ13501.1 | |||
| AK026362.1 | BAB15460.1 | |||
| BC004121.2 | AAH04121.1 | |||
| BC029361.1 | AAH29361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470129 | Chr.19:17188084 - 17341718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map