Assay Details
Target Gene Details
Entrez Gene ID: | 139221 |
Gene Name: | MUM1 like 1 |
Gene Aliases: |
- |
Location: |
Chr.X:106168278-106208961 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MUM1L1 | NM_001171020.1 | 4 | 2309 | NP_001164491.1 |
| NM_152423.4 | 5 | 2445 | NP_689636.3 | |
| XM_005262077.2 | 4 | 2226 | XP_005262134.1 | |
| XM_011530856.1 | 4 | 2303 | XP_011529158.1 | |
| XM_011530857.1 | 4 | 2272 | XP_011529159.1 | |
| XM_017029269.1 | 5 | 2439 | XP_016884758.1 | |
| AK056478.1 | 3 | 2095 | BAB71194.1 | |
| AK090835.1 | 1 | 961 | BAC03529.1 | |
| BC031229.1 | 4 | 2309 | AAH31229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752312 | Chr.X:105705935 - 107123872 on Build GRCh38 | Gain |
|
| nsv521259 | Chr.X:105935140 - 106434204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map