Assay Details
Target Gene Details
Entrez Gene ID: | 23314 |
Gene Name: | SATB homeobox 2 |
Gene Aliases: |
GLSS |
Location: |
Chr.2:199269500-199471266 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SATB2 | NM_001172509.1 | 11 | 3944 | NP_001165980.1 |
| NM_001172517.1 | 12 | 3540 | NP_001165988.1 | |
| NM_015265.3 | 12 | 3520 | NP_056080.1 | |
| XM_005246396.2 | 10 | 3175 | XP_005246453.1 | |
| XM_017003656.1 | 11 | 3175 | XP_016859145.1 | |
| AB028957.1 | 11 | 3213 | ||
| AB209376.1 | 12 | 3540 | BAD92613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3104 | Chr.2:199264502 - 199299835 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map