Assay Details
Target Gene Details
Entrez Gene ID: | 57449 |
Gene Name: | pleckstrin homology and RhoGEF domain containing G5 |
Gene Aliases: |
CMTRIC, DSMA4, GEF720, Syx, Tech |
Location: |
Chr.1:6466092-6520061 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHG5 | NM_001042663.1 | NP_001036128.1 | ||
| NM_001042664.1 | NP_001036129.1 | |||
| NM_001042665.1 | NP_001036130.1 | |||
| NM_001265592.1 | NP_001252521.1 | |||
| NM_001265593.1 | NP_001252522.1 | |||
| NM_001265594.1 | NP_001252523.1 | |||
| NM_020631.4 | NP_065682.2 | |||
| NM_198681.3 | NP_941374.2 | |||
| AB018263.2 | ||||
| AB097001.1 | BAC77354.1 | |||
| AK024676.1 | ||||
| AK096347.1 | ||||
| AK125550.1 | ||||
| AK131074.1 | BAC85124.1 | |||
| AK294875.1 | ||||
| AK299523.1 | ||||
| AK301239.1 | ||||
| BC015231.1 | AAH15231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009548 | Chr.1:6342976 - 6517352 on Build GRCh38 | Loss |
|
| dgv80n54 | Chr.1:6396412 - 6512115 on Build GRCh38 | Loss |
|
| nsv545267 | Chr.1:6466264 - 6504096 on Build GRCh38 | Loss |
|
| nsv954048 | Chr.1:6414241 - 6500740 on Build GRCh38 | Deletion |
|
| nsv470690 | Chr.1:6434339 - 6512115 on Build GRCh38 | Gain |
|
| nsv7880 | Chr.1:6417968 - 6611914 on Build GRCh38 | Gain+Loss |
|
| nsv545264 | Chr.1:6457119 - 6492439 on Build GRCh38 | Loss |
|
| nsv545240 | Chr.1:6351154 - 6512115 on Build GRCh38 | Loss |
|
| nsv470691 | Chr.1:6460251 - 6517705 on Build GRCh38 | Loss |
|
| nsv462061 | Chr.1:6463370 - 6520836 on Build GRCh38 | Loss |
|
| nsv831536 | Chr.1:6429326 - 6605343 on Build GRCh38 | Loss |
|
| nsv508836 | Chr.1:6406896 - 6501029 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76386795] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map