Assay Details
Target Gene Details
Entrez Gene ID: | 23620 |
Gene Name: | neurotensin receptor 2 |
Gene Aliases: |
NTR2 |
Location: |
Chr.2:11658178-11670206 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTSR2 | NM_012344.3 | 4 | 1153 | NP_036476.1 |
| XM_005246156.3 | 3 | 882 | XP_005246213.1 | |
| XM_006711876.3 | 4 | 1098 | XP_006711939.1 | |
| XM_006711877.3 | 4 | 1089 | XP_006711940.1 | |
| XM_017003738.1 | 4 | 1149 | XP_016859227.1 | |
| AY429107.1 | 4 | 1079 | AAR07902.1 | |
| BC022501.1 | 4 | 1114 | AAH22501.1 | |
| BC037776.2 | 5 | 1067 | AAH37776.1 | |
| Y10148.1 | 4 | 1115 | CAA71233.1 |
More Information
Set Membership: |
|

Genomic Map