Assay Details
Target Gene Details
Entrez Gene ID: | 311 |
Gene Name: | annexin A11 |
Gene Aliases: |
ANX11, CAP50 |
Location: |
Chr.10:80153953-80205677 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANXA11 | NM_001157.2 | NP_001148.1 | ||
| NM_001278407.1 | NP_001265336.1 | |||
| NM_001278408.1 | NP_001265337.1 | |||
| NM_001278409.1 | NP_001265338.1 | |||
| NM_145868.1 | NP_665875.1 | |||
| NM_145869.1 | NP_665876.1 | |||
| XM_005269741.4 | 1 | 364 | XP_005269798.1 | |
| XM_005269742.1 | XP_005269799.1 | |||
| XM_006717813.1 | XP_006717876.1 | |||
| XM_006717814.3 | XP_006717877.1 | |||
| XM_011539735.1 | XP_011538037.1 | |||
| XM_011539736.2 | XP_011538038.1 | |||
| AB209770.1 | BAD93007.1 | |||
| AJ278463.1 | CAB94995.1 | |||
| AJ278464.1 | CAB94996.1 | |||
| AJ278465.1 | CAB94997.1 | |||
| AK000567.1 | ||||
| AK056107.1 | ||||
| AK126287.1 | ||||
| AK222918.1 | BAD96638.1 | |||
| AK298362.1 | ||||
| AK300509.1 | 1 | 399 | ||
| AK301047.1 | ||||
| BC007564.2 | AAH07564.1 | |||
| BT019934.1 | AAV38737.1 | |||
| CR450323.1 | CAG29319.1 | |||
| DA558997.1 | ||||
| L19605.1 | AAA19734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551635 | Chr.10:79925268 - 80212630 on Build GRCh38 | Loss |
|
| esv3623946 | Chr.10:80133465 - 80175734 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map