Assay Details
Target Gene Details
Entrez Gene ID: | 1069 |
Gene Name: | centrin 2 |
Gene Aliases: |
CALT, CEN2 |
Location: |
Chr.X:152827327-152830757 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CETN2 | NM_004344.1 | 5 | 850 | NP_004335.1 |
| BC005334.1 | 5 | 850 | AAH05334.1 | |
| BC013873.2 | 5 | 848 | AAH13873.1 | |
| X72964.1 | 5 | 850 | CAA51467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758901 | Chr.X:152745001 - 152849208 on Build GRCh38 | Gain |
|
| esv25868 | Chr.X:152816977 - 152847832 on Build GRCh38 | Gain |
|
| nsv470373 | Chr.X:152796870 - 152935948 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map