Assay Details
Target Gene Details
Entrez Gene ID: | 7283 |
Gene Name: | tubulin gamma 1 |
Gene Aliases: |
CDCBM4, GCP-1, TUBG, TUBGCP1 |
Location: |
Chr.17:42609340-42615238 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBG1 | NM_001070.4 | NP_001061.2 | ||
| BC000619.2 | AAH00619.1 | |||
| BG167819.1 | ||||
| M61764.1 | AAA52620.1 |
More Information
Set Membership: |
|

Genomic Map