Assay Details
Target Gene Details
Entrez Gene ID: | 64344 |
Gene Name: | hypoxia inducible factor 3 alpha subunit |
Gene Aliases: |
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17 |
Location: |
Chr.19:46297046-46343433 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIF3A | NM_022462.4 | 1 | 54 | NP_071907.4 |
| NM_152795.3 | NP_690008.2 | |||
| NM_152796.2 | 1 | 54 | NP_690009.1 | |
| XM_005259152.4 | 2 | 100 | XP_005259209.1 | |
| XM_005259155.4 | 2 | 100 | XP_005259212.1 | |
| XM_005259156.4 | 2 | 100 | XP_005259213.1 | |
| XM_017027132.1 | 2 | 100 | XP_016882621.1 | |
| XM_017027133.1 | 2 | 100 | XP_016882622.1 | |
| XM_017027134.1 | XP_016882623.1 | |||
| XM_017027136.1 | 2 | 100 | XP_016882625.1 | |
| XM_017027138.1 | XP_016882627.1 | |||
| XM_017027139.1 | XP_016882628.1 | |||
| XM_017027140.1 | XP_016882629.1 | |||
| XM_017027142.1 | XP_016882631.1 | |||
| AB118749.1 | BAD93355.1 | |||
| AB295039.1 | ||||
| AF463492.1 | AAL69947.1 | |||
| AK021421.1 | BAB13819.1 | |||
| AK021653.1 | BAB13865.1 | |||
| AK024095.1 | BAB14824.1 | |||
| AK027725.1 | BAB55324.1 | |||
| AK297828.1 | ||||
| AK304747.1 | ||||
| BC026308.1 | AAH26308.1 | |||
| BC080551.1 | AAH80551.1 | |||
| BI772109.1 | ||||
| DA559979.1 | ||||
| DB176660.1 | ||||
| DC423791.1 | 1 | 54 | ||
| DN989983.1 | 1 | 139 | ||
| DR001068.1 |
More Information
Set Membership: |
|

Genomic Map