Assay Details
Target Gene Details
Entrez Gene ID: | 85291 |
Gene Name: | keratin associated protein 4-2 |
Gene Aliases: |
KAP4.2, KRTAP4.2 |
Location: |
Chr.17:41177446-41178208 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP4-2 | NM_033062.3 | 1 | 529 | NP_149051.1 |
| AJ406934.1 | 1 | 529 | CAC27573.1 | |
| BC127252.1 | 1 | 509 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833446 | Chr.17:41059613 - 41247007 on Build GRCh38 | Gain |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map