Assay Details
Target Gene Details
Entrez Gene ID: | 8548 |
Gene Name: | basic leucine zipper nuclear factor 1 |
Gene Aliases: |
GOLGIN-45, JEM-1, JEM-1s, JEM1 |
Location: |
Chr.1:169367879-169396544 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLZF1 | NM_001320973.1 | NP_001307902.1 | ||
| NM_003666.3 | NP_003657.1 | |||
| XM_005245561.4 | XP_005245618.1 | |||
| XM_011510092.2 | XP_011508394.1 | |||
| XM_017002645.1 | XP_016858134.1 | |||
| AK223533.1 | BAD97253.1 | |||
| AK291725.1 | ||||
| AK307151.1 | ||||
| BC020716.1 | AAH20716.1 | |||
| U79751.1 | AAB63110.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv22143 | Chr.1:169090390 - 169652030 on Build GRCh38 | Gain+Loss |
|
| nsv831871 | Chr.1:169351435 - 169517442 on Build GRCh38 | Loss |
|
| esv3890760 | Chr.1:169289068 - 169388040 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map