Assay Details
Target Gene Details
Entrez Gene ID: | 64834 |
Gene Name: | ELOVL fatty acid elongase 1 |
Gene Aliases: |
CGI-88, Ssc1 |
Location: |
Chr.1:43363397-43368074 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL1 | NM_001256399.1 | 8 | 1091 | NP_001243328.1 |
| NM_001256401.1 | 7 | 917 | NP_001243330.1 | |
| NM_001256402.1 | 7 | 807 | NP_001243331.1 | |
| NM_022821.3 | 8 | 998 | NP_073732.1 | |
| NR_046117.1 | 8 | 925 | ||
| AF151846.1 | 8 | 941 | AAD34083.1 | |
| AF336793.1 | 8 | 927 | AAL71993.1 | |
| AK001653.1 | 8 | 927 | BAA91813.1 | |
| AK130174.1 | 8 | 901 | ||
| AK222498.1 | 8 | 943 | BAD96218.1 | |
| AK298163.1 | 7 | 854 | ||
| BC000618.2 | 8 | 919 | AAH00618.1 | |
| BC095456.1 | 8 | 858 | AAH95456.1 | |
| BM556982.1 | 7 | 746 | ||
| BU180063.1 | 1 | 130 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map