Assay Details
Target Gene Details
Entrez Gene ID: | 5393 |
Gene Name: | exosome component 9 |
Gene Aliases: |
PM/Scl-75, PMSCL1, RRP45, Rrp45p, p5, p6 |
Location: |
Chr.4:121801295-121817021 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOSC9 | NM_001034194.1 | 10 | 1181 | NP_001029366.1 |
| NM_005033.2 | 10 | 1181 | NP_005024.2 | |
| XM_011532035.2 | 10 | 1203 | XP_011530337.1 | |
| AJ505989.1 | 10 | 1178 | CAD44530.1 | |
| AJ517294.1 | 10 | 1178 | CAD56889.1 | |
| AK301967.1 | 10 | 1180 | ||
| AK307548.1 | 10 | 1120 | ||
| AK310894.1 | 5 | 541 | ||
| BC142978.1 | 10 | 1152 | ||
| BU615347.1 | 4 | 481 | ||
| BX649106.1 | 10 | 1189 | ||
| M58460.1 | 9 | 1240 | AAA58384.1 | |
| U09215.1 | 8 | 821 | AAA18832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997539 | Chr.4:121622949 - 122353111 on Build GRCh38 | Gain |
|
| esv3602016 | Chr.4:121811764 - 121818497 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map