Assay Details
Target Gene Details
Entrez Gene ID: | 4776 |
Gene Name: | nuclear factor of activated T-cells 4 |
Gene Aliases: |
NF-AT3, NF-ATC4, NFAT3 |
Location: |
Chr.14:24366911-24379604 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFATC4 | NM_001136022.2 | 10 | 4183 | NP_001129494.1 |
| NM_001198965.1 | 10 | 3088 | NP_001185894.1 | |
| NM_001198966.2 | 10 | 3066 | NP_001185895.1 | |
| NM_001198967.2 | 11 | 3100 | NP_001185896.1 | |
| NM_001288802.1 | 9 | 3974 | NP_001275731.1 | |
| NM_001320043.1 | 11 | 3424 | NP_001306972.1 | |
| NM_004554.4 | 10 | 3412 | NP_004545.2 | |
| XM_011536797.2 | XP_011535099.1 | |||
| XM_011536798.1 | XP_011535100.1 | |||
| XM_011536799.2 | XP_011535101.1 | |||
| AK293185.1 | ||||
| BC008857.2 | AAH08857.2 | |||
| BC053855.1 | 10 | 3228 | AAH53855.1 | |
| EU887634.1 | 10 | 4155 | ||
| EU887637.1 | 10 | 3971 | ||
| EU887640.1 | 10 | 4024 | ||
| EU887643.1 | 9 | 3850 | ||
| EU887646.1 | 9 | 3974 | ||
| EU887649.1 | 8 | 4096 | ||
| EU887652.1 | 6 | 2847 |
More Information
Set Membership: |
|

Genomic Map