Assay Details
Target Gene Details
Entrez Gene ID: | 1361 |
Gene Name: | carboxypeptidase B2 |
Gene Aliases: |
CPU, PCPB, TAFI |
Location: |
Chr.13:46053186-46105076 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPB2 | NM_001278541.1 | 10 | 1349 | NP_001265470.1 |
| NM_001872.4 | 11 | 1460 | NP_001863.3 | |
| XM_017020393.1 | 11 | 1403 | XP_016875882.1 | |
| AA700377.1 | 1 | 300 | ||
| AB011969.1 | 11 | 1249 | BAA90475.1 | |
| AK290829.1 | 11 | 1408 | ||
| AV719308.1 | 1 | 307 | ||
| BC007057.1 | 11 | 1417 | AAH07057.1 | |
| M75106.1 | 11 | 1412 | AAA60042.1 |
Target Gene Details
Entrez Gene ID: | 100509894 |
Gene Name: | CPB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:46052848-46101347 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPB2-AS1 | NR_046226.1 | |||
| NR_046227.1 | ||||
| AK095119.1 | ||||
| AK124928.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv223e201 | Chr.13:45377870 - 46464067 on Build GRCh38 | Deletion |
|
| esv2673081 | Chr.13:46029727 - 46073598 on Build GRCh38 | Deletion |
|
| nsv832598 | Chr.13:45974228 - 46149047 on Build GRCh38 | Gain |
|
| nsv455876 | Chr.13:46044155 - 46739284 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map