Assay Details
Target Gene Details
Entrez Gene ID: | 283160 |
Gene Name: | olfactory receptor family 8 subfamily D member 2 (gene/pseudogene) |
Gene Aliases: |
JCG2 |
Location: |
Chr.11:124319262-124320197 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR8D2 | NM_001002918.1 | 1 | 836 | NP_001002918.1 |
| AF162668.1 | 1 | 836 | AAG43386.1 | |
| BC137043.1 | 1 | 927 | ||
| BC137045.1 | 1 | 927 |
Target Gene Details
Entrez Gene ID: | 219865 |
Gene Name: | olfactory receptor family 8 subfamily G member 5 |
Gene Aliases: |
OR11-298, OR8G5P, OR8G6 |
Location: |
Chr.11:124241158-124323283 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR8G5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826115 | Chr.11:124319324 - 124320385 on Build GRCh38 | Loss |
|
| nsv826113 | Chr.11:124267376 - 124372263 on Build GRCh38 | Loss |
|
| esv3627975 | Chr.11:124318013 - 124328419 on Build GRCh38 | Loss |
|
| esv3892105 | Chr.11:124309052 - 124530715 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map