Assay Details
Target Gene Details
Entrez Gene ID: | 54943 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C28 |
Gene Aliases: |
C21orf55, C21orf78 |
Location: |
Chr.21:33488055-33491723 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC28 | NM_001040192.2 | NP_001035282.1 | ||
| NM_001320746.1 | NP_001307675.1 | |||
| NM_017833.4 | 1 | 104 | NP_060303.2 | |
| AF462153.2 | 1 | 94 | AAL59618.2 | |
| AK000468.1 | 1 | 94 | BAA91185.1 | |
| AK225514.1 | 1 | 94 | ||
| DA234338.1 | ||||
| DA249316.1 | 1 | 104 | ||
| HY130099.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509795 | Chr.21:33457276 - 33521074 on Build GRCh38 | Insertion |
|
| nsv524420 | Chr.21:33455533 - 33491977 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116108104] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map