Assay Details
Target Gene Details
Entrez Gene ID: | 159 |
Gene Name: | adenylosuccinate synthase |
Gene Aliases: |
ADEH, ADSS 2 |
Location: |
Chr.1:244408492-244452134 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADSS | NM_001126.3 | NP_001117.2 | ||
| XM_011544108.2 | XP_011542410.1 | |||
| AK025514.1 | ||||
| AK303886.1 | ||||
| BC012356.1 | AAH12356.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516557 | Chr.1:244381516 - 244538198 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map