Assay Details
Target Gene Details
Entrez Gene ID: | 23061 |
Gene Name: | TBC1 domain family member 9B |
Gene Aliases: |
GRAMD9B |
Location: |
Chr.5:179862071-179907856 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D9B | NM_015043.3 | 21 | 4752 | NP_055858.2 |
| NM_198868.2 | 22 | 4803 | NP_942568.2 | |
| AK001196.1 | 8 | 2365 | ||
| AK001473.1 | 3 | 1827 | ||
| AK026096.1 | 1 | 1314 | ||
| BC000701.2 | 8 | 2386 | AAH00701.1 | |
| BC008919.2 | 10 | 2734 | AAH08919.3 | |
| BC080659.1 | 10 | 2813 | AAH80659.1 | |
| BX641107.1 | 2 | 3469 | CAE46050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469579 | Chr.5:179757830 - 179933191 on Build GRCh38 | Loss |
|
| esv3607755 | Chr.5:179853436 - 179869950 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map