Assay Details
Target Gene Details
Entrez Gene ID: | 57060 |
Gene Name: | poly(rC) binding protein 4 |
Gene Aliases: |
CBP, LIP4, MCG10 |
Location: |
Chr.3:51957454-51967466 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCBP4 | NM_001174100.1 | NP_001167571.1 | ||
| NM_020418.3 | NP_065151.2 | |||
| NM_033008.2 | NP_127501.1 | |||
| NM_033010.2 | NP_127503.1 | |||
| XM_005265330.2 | XP_005265387.1 | |||
| XM_005265331.2 | XP_005265388.1 | |||
| XM_005265332.2 | XP_005265389.1 | |||
| XM_006713268.2 | XP_006713331.1 | |||
| XM_006713269.1 | XP_006713332.1 | |||
| XM_006713270.1 | XP_006713333.1 | |||
| XM_006713271.2 | XP_006713334.1 | |||
| XM_017006916.1 | XP_016862405.1 | |||
| XM_017006917.1 | XP_016862406.1 | |||
| XM_017006918.1 | XP_016862407.1 | |||
| XM_017006919.1 | XP_016862408.1 | |||
| XM_017006920.1 | XP_016862409.1 | |||
| AF092441.1 | AAP97216.1 | |||
| AF176330.1 | AAG09241.1 | |||
| AF257770.1 | 3 | 602 | AAG02184.1 | |
| AF257771.1 | 3 | 602 | AAG02185.1 | |
| AK001244.1 | ||||
| AK023993.1 | BAB14761.1 | |||
| AK225815.1 | ||||
| BC003008.1 | AAH03008.1 | |||
| BC004153.2 | AAH04153.1 | |||
| BC017098.1 | AAH17098.1 | |||
| BX647811.1 | 4 | 717 | ||
| DA701799.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003453 | Chr.3:51928065 - 52065425 on Build GRCh38 | Gain |
|
| nsv508922 | Chr.3:51940932 - 52009520 on Build GRCh38 | Insertion |
|
| nsv954857 | Chr.3:51932485 - 52056584 on Build GRCh38 | Deletion |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map