Assay Details
Target Gene Details
Entrez Gene ID: | 10922 |
Gene Name: | Fas activated serine/threonine kinase |
Gene Aliases: |
FAST |
Location: |
Chr.7:151076621-151080883 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FASTK | NM_001258461.1 | NP_001245390.1 | ||
| NM_006712.4 | NP_006703.1 | |||
| NM_033015.3 | NP_148936.2 | |||
| XM_005249932.1 | XP_005249989.1 | |||
| XM_005249933.3 | XP_005249990.1 | |||
| XM_011515761.2 | XP_011514063.1 | |||
| XM_011515762.2 | XP_011514064.1 | |||
| XM_011515763.1 | XP_011514065.1 | |||
| XM_017011704.1 | XP_016867193.1 | |||
| XM_017011705.1 | XP_016867194.1 | |||
| XM_017011706.1 | XP_016867195.1 | |||
| AK023141.1 | BAB14425.1 | |||
| AK223132.1 | BAD96852.1 | |||
| BC000377.2 | AAH00377.1 | |||
| BC011770.2 | AAH11770.1 | |||
| BC039026.2 | AAH39026.1 | |||
| BM799420.1 | ||||
| DA253544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv11724n54 | Chr.7:151054503 - 151135826 on Build GRCh38 | Loss |
|
| nsv831188 | Chr.7:151038117 - 151215789 on Build GRCh38 | Loss |
|
| nsv951648 | Chr.7:150931413 - 151106413 on Build GRCh38 | Deletion |
|
| nsv465215 | Chr.7:151065784 - 151128000 on Build GRCh38 | Loss |
|
| dgv6756n100 | Chr.7:151033912 - 151093430 on Build GRCh38 | Gain |
|
| nsv6004 | Chr.7:151067919 - 151098858 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map