Assay Details
Target Gene Details
Entrez Gene ID: | 7415 |
Gene Name: | valosin containing protein |
Gene Aliases: |
ALS14, CMT2Y, HEL-220, HEL-S-70, IBMPFD, IBMPFD1, TERA, p97 |
Location: |
Chr.9:35056068-35072742 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VCP | NM_007126.3 | 17 | 3071 | NP_009057.1 |
| AF100752.1 | AAD43016.1 | |||
| AL137377.1 | 8 | 1557 | CAB70717.1 | |
| BC007562.1 | 13 | 2197 | AAH07562.2 | |
| BC110913.1 | 17 | 2881 | AAI10914.1 | |
| BC121794.1 | ||||
| BC122550.1 | ||||
| FJ224344.1 | 17 | 2894 | ||
| FJ224352.1 |
More Information
Set Membership: |
|

Genomic Map