Assay Details
Target Gene Details
Entrez Gene ID: | 9221 |
Gene Name: | nucleolar and coiled-body phosphoprotein 1 |
Gene Aliases: |
NOPP130, NOPP140, NS5ATP13, P130 |
Location: |
Chr.10:102152176-102163871 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOLC1 | NM_001284388.1 | 5 | 766 | NP_001271317.1 |
| NM_001284389.1 | 5 | 769 | NP_001271318.1 | |
| NM_004741.4 | 5 | 766 | NP_004732.2 | |
| XM_005270273.1 | 5 | 743 | XP_005270330.1 | |
| AI937215.1 | 3 | 302 | ||
| AK056755.1 | 4 | 389 | ||
| AK225101.1 | 5 | 556 | ||
| AK314363.1 | 5 | 553 | ||
| AY820769.1 | 5 | 534 | AAV67777.1 | |
| BC001883.1 | 5 | 541 | AAH01883.1 | |
| BC006769.2 | AAH06769.1 | |||
| D21262.1 | 5 | 527 | BAA04803.1 | |
| Z34289.1 | 5 | 585 | CAA84063.1 |
More Information
Set Membership: |
|

Genomic Map