Assay Details
Target Gene Details
Entrez Gene ID: | 100529262 |
Gene Name: | MIA-RAB4B readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.19:40775537-40796944 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIA-RAB4B | NR_037775.1 | |||
| AF087861.1 | AAP97171.1 | |||
| AK302147.1 |
Target Gene Details
Entrez Gene ID: | 53916 |
Gene Name: | RAB4B, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.19:40778219-40796944 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB4B | NM_016154.4 | NP_057238.3 | ||
| AF165522.1 | AAD45923.1 | |||
| AF217985.1 | AAG17228.1 | |||
| AF498935.1 | AAM21083.1 | |||
| BC033081.1 | AAH33081.1 | |||
| BC046927.1 | AAH46927.1 | |||
| BG706803.1 |
Target Gene Details
Entrez Gene ID: | 100529264 |
Gene Name: | RAB4B-EGLN2 readthrough (NMD candidate) |
Gene Aliases: |
RERT-lncRNA |
Location: |
Chr.19:40778219-40808441 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB4B-EGLN2 | NR_037791.1 | |||
| AK291385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833832 | Chr.19:40754458 - 40828692 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map