Assay Details
Target Gene Details
Entrez Gene ID: | 57198 |
Gene Name: | ATPase phospholipid transporting 8B2 |
Gene Aliases: |
ATPID |
Location: |
Chr.1:154325541-154351307 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP8B2 | NM_020452.3 | 13 | 1202 | NP_065185.1 |
| XM_005245355.2 | 12 | 1349 | XP_005245412.1 | |
| XM_005245356.3 | 13 | 1302 | XP_005245413.1 | |
| XM_017001871.1 | 12 | 1349 | XP_016857360.1 | |
| XM_017001872.1 | 13 | 1298 | XP_016857361.1 | |
| AB032963.1 | 3 | 334 | BAA86451.1 | |
| AK054886.1 | 13 | 1356 | BAB70822.1 | |
| AY302537.1 | 13 | 1202 | AAQ19027.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521887 | Chr.1:154264094 - 154711630 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map