Assay Details
Target Gene Details
Entrez Gene ID: | 254240 |
Gene Name: | BPI fold containing family C |
Gene Aliases: |
BPIL2 |
Location: |
Chr.22:32413845-32464465 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPIFC | NM_174932.2 | NP_777592.1 | ||
| XM_011530088.2 | XP_011528390.1 | |||
| XM_011530089.1 | XP_011528391.1 | |||
| XM_011530090.1 | XP_011528392.1 | |||
| XM_011530091.2 | XP_011528393.1 | |||
| XM_017028740.1 | XP_016884229.1 | |||
| AF465766.1 | AAM73984.1 | |||
| AK301685.1 | ||||
| BC131582.1 | ||||
| CU456409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066118 | Chr.22:32383091 - 32419347 on Build GRCh38 | Gain |
|
| nsv459874 | Chr.22:32178287 - 32470947 on Build GRCh38 | Gain |
|
| nsv1055701 | Chr.22:32410608 - 32449416 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map