Assay Details
Target Gene Details
Entrez Gene ID: | 343521 |
Gene Name: | Tctex1 domain containing 4 |
Gene Aliases: |
- |
Location: |
Chr.1:44805908-44807456 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCTEX1D4 | NM_001013632.3 | 2 | 1113 | NP_001013654.1 |
| XM_006710614.3 | 3 | 798 | XP_006710677.1 | |
| XM_011541375.2 | 3 | 848 | XP_011539677.1 | |
| XM_011541376.2 | 3 | 704 | XP_011539678.1 | |
| XM_017001187.1 | 2 | 747 | XP_016856676.1 | |
| BC092499.1 | 2 | 1113 | AAH92499.1 | |
| DQ132441.1 | 2 | 1113 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950705 | Chr.1:44801029 - 44813128 on Build GRCh38 | Deletion |
|
| nsv546145 | Chr.1:44752870 - 44827846 on Build GRCh38 | Loss |
|
| nsv546146 | Chr.1:44792469 - 44853080 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map