Assay Details
Target Gene Details
Entrez Gene ID: | 3992 |
Gene Name: | fatty acid desaturase 1 |
Gene Aliases: |
D5D, FADS6, FADSD5, LLCDL1, TU12 |
Location: |
Chr.11:61799625-61817057 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FADS1 | NM_013402.4 | 12 | 3407 | NP_037534.3 |
| XM_011545022.2 | 12 | 3081 | XP_011543324.1 | |
| AF035284.1 | 1 | 1346 | ||
| AF084558.1 | 12 | 3186 | AAG23120.1 | |
| AF271778.1 | 1 | 260 | AAG44789.1 | |
| AL512760.1 | 12 | 3188 | CAC21679.1 | |
| AL834479.1 | 4 | 2050 | CAD39138.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528697 | Chr.11:61690385 - 61919561 on Build GRCh38 | Loss |
|
| nsv343 | Chr.11:61770698 - 61802486 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map