Assay Details
Target Gene Details
Entrez Gene ID: | 1128 |
Gene Name: | cholinergic receptor muscarinic 1 |
Gene Aliases: |
HM1, M1, M1R |
Location: |
Chr.11:62908675-62921861 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRM1 | NM_000738.2 | 2 | 1684 | NP_000729.2 |
| XM_011544742.2 | 2 | 1385 | XP_011543044.1 | |
| AF385587.1 | 1 | 1293 | AAK68112.1 | |
| AF498915.1 | 1 | 1283 | AAM18938.1 | |
| AK312374.1 | 2 | 1444 | ||
| BC007740.2 | 2 | 1405 | AAH07740.1 | |
| BC022984.1 | 2 | 1684 | AAH22984.1 | |
| BT007166.1 | 1 | 1283 | AAP35830.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34011 | Chr.11:62885307 - 63226230 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map