Assay Details
Target Gene Details
Entrez Gene ID: | 9070 |
Gene Name: | ASH2 like histone lysine methyltransferase complex subunit |
Gene Aliases: |
ASH2, ASH2L1, ASH2L2, Bre2 |
Location: |
Chr.8:38104965-38140080 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASH2L | NM_001105214.2 | NP_001098684.1 | ||
| NM_001261832.1 | NP_001248761.1 | |||
| NM_001282272.1 | NP_001269201.1 | |||
| NM_004674.4 | NP_004665.2 | |||
| XM_005273682.1 | XP_005273739.1 | |||
| XM_005273683.1 | XP_005273740.1 | |||
| XM_006716412.1 | XP_006716475.1 | |||
| XM_006716413.2 | XP_006716476.1 | |||
| AB020982.1 | BAA35127.1 | |||
| AF056717.1 | AAC13563.1 | |||
| AF056718.1 | AAC13564.1 | |||
| AK291750.1 | ||||
| AK291938.1 | ||||
| AK293699.1 | ||||
| AK298483.1 | ||||
| AK311336.1 | ||||
| BC015936.2 | AAH15936.1 | |||
| DC326351.1 | ||||
| DC353644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616918 | Chr.8:38080740 - 38119204 on Build GRCh38 | Gain |
|
| esv3616917 | Chr.8:38078635 - 38160673 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map