Assay Details
Target Gene Details
Entrez Gene ID: | 221060 |
Gene Name: | chromosome 10 open reading frame 111 |
Gene Aliases: |
bA455B2.4 |
Location: |
Chr.10:15095385-15097319 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf111 | NM_153244.1 | 2 | 1419 | NP_694976.1 |
| BC029034.1 | 2 | 1419 | AAH29034.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831794 | Chr.10:14993658 - 15148891 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73602469] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map