Assay Details
Target Gene Details
Entrez Gene ID: | 6570 |
Gene Name: | solute carrier family 18 member A1 |
Gene Aliases: |
CGAT, VAT1, VMAT1 |
Location: |
Chr.8:20144855-20183206 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC18A1 | NM_001135691.2 | NP_001129163.1 | ||
| NM_001142324.1 | NP_001135796.1 | |||
| NM_001142325.1 | NP_001135797.1 | |||
| NM_003053.3 | NP_003044.1 | |||
| XM_011544625.1 | XP_011542927.1 | |||
| XM_011544626.1 | XP_011542928.1 | |||
| XM_011544627.1 | XP_011542929.1 | |||
| AK313720.1 | ||||
| BC006317.2 | AAH06317.1 | |||
| BC009387.2 | AAH09387.1 | |||
| U39905.1 | AAC50472.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv438045 | Chr.8:20135591 - 20165660 on Build GRCh38 | Loss |
|
| nsv1030738 | Chr.8:20131683 - 20164639 on Build GRCh38 | Gain |
|
| nsv508498 | Chr.8:20127378 - 20174629 on Build GRCh38 | Deletion |
|
| esv3891373 | Chr.8:20127871 - 20164378 on Build GRCh38 | Loss |
|
| esv3616559 | Chr.8:20127120 - 20166631 on Build GRCh38 | Loss |
|
| nsv824563 | Chr.8:20127252 - 20168579 on Build GRCh38 | Loss |
|
| nsv610759 | Chr.8:19704486 - 20211331 on Build GRCh38 | Gain |
|
| nsv522185 | Chr.8:19097292 - 20497802 on Build GRCh38 | Gain |
|
| esv2759602 | Chr.8:20129060 - 20172092 on Build GRCh38 | Loss |
|
| nsv1033511 | Chr.8:19893788 - 20767580 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map