Assay Details
Target Gene Details
Entrez Gene ID: | 54982 |
Gene Name: | ceroid-lipofuscinosis, neuronal 6, late infantile, variant |
Gene Aliases: |
CLN4A, HsT18960, nclf |
Location: |
Chr.15:68206992-68229742 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLN6 | NM_017882.2 | 7 | 2152 | NP_060352.1 |
| AK000568.1 | 7 | 2097 | BAA91260.1 | |
| AK027604.1 | 7 | 2120 | BAB55226.1 | |
| AK074753.1 | 4 | 1646 | BAC11181.1 | |
| BC010849.1 | 7 | 2066 | AAH10849.1 | |
| BC013130.1 | 7 | 2003 | AAH13130.1 | |
| CA446197.1 | 1 | 107 |
More Information
Additional Information:
For this assay, SNP(s) [rs141489068] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map