Assay Details
Target Gene Details
Entrez Gene ID: | 50614 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 9 |
Gene Aliases: |
GALNAC-T9, GALNACT9 |
Location: |
Chr.12:132195829-132329364 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT9 | NM_001122636.1 | 7 | 1273 | NP_001116108.1 |
| NM_021808.3 | 3 | 337 | NP_068580.2 | |
| XM_017019371.1 | 6 | 1005 | XP_016874860.1 | |
| XM_017019372.1 | 6 | 965 | XP_016874861.1 | |
| XM_017019373.1 | 7 | 1322 | XP_016874862.1 | |
| XM_017019374.1 | 6 | 942 | XP_016874863.1 | |
| AB040672.2 | 7 | 1157 | BAB13699.2 | |
| AF458594.1 | 3 | 334 | AAM49722.1 | |
| AK054820.1 | 4 | 474 | ||
| AK055773.1 | 6 | 930 | ||
| BC093817.1 | 2 | 147 | ||
| BC093819.1 | 2 | 147 | ||
| BI760629.1 | 2 | 409 | ||
| CB154648.1 | ||||
| DR001000.1 | 2 | 141 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3019n54 | Chr.12:132181784 - 132203592 on Build GRCh38 | Loss |
|
| nsv560966 | Chr.12:132194518 - 132483313 on Build GRCh38 | Gain |
|
| esv3631288 | Chr.12:132190709 - 132205907 on Build GRCh38 | Gain |
|
| esv3580551 | Chr.12:132196503 - 132206310 on Build GRCh38 | Loss |
|
| nsv952512 | Chr.12:132192256 - 132711214 on Build GRCh38 | Deletion |
|
| nsv520897 | Chr.12:132199317 - 132218324 on Build GRCh38 | Loss |
|
| nsv1041402 | Chr.12:132076412 - 132441297 on Build GRCh38 | Gain |
|
| nsv528119 | Chr.12:132174775 - 132218324 on Build GRCh38 | Loss |
|
| nsv826563 | Chr.12:132125383 - 132622340 on Build GRCh38 | Gain |
|
| dgv103n21 | Chr.12:132117051 - 132442096 on Build GRCh38 | Loss |
|
| nsv428285 | Chr.12:132065104 - 132208513 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117160424] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map