Assay Details
Target Gene Details
Entrez Gene ID: | 3431 |
Gene Name: | SP110 nuclear body protein |
Gene Aliases: |
IFI41, IFI75, IPR1, VODI |
Location: |
Chr.2:230165466-230225729 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP110 | NM_004509.3 | 18 | 2386 | NP_004500.3 |
| NM_080424.2 | 19 | 2458 | NP_536349.2 | |
| XM_005246525.3 | 20 | 2494 | XP_005246582.1 | |
| XM_006712487.3 | 19 | 2422 | XP_006712550.1 | |
| XM_006712489.3 | 18 | 2281 | XP_006712552.1 | |
| XM_011511088.2 | 20 | 2572 | XP_011509390.1 | |
| XM_011511089.2 | 19 | 2427 | XP_011509391.1 | |
| XM_011511090.2 | 19 | 2422 | XP_011509392.1 | |
| XM_011511091.2 | 19 | 2353 | XP_011509393.1 | |
| XM_011511092.2 | 18 | 2265 | XP_011509394.1 | |
| XM_017003968.1 | 21 | 2647 | XP_016859457.1 | |
| AF280095.1 | 18 | 2223 | AAG09826.1 | |
| AK026488.1 | 19 | 2295 | ||
| AK128274.1 | 3 | 970 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3594596 | Chr.2:230072135 - 230170374 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map